Di-fmoc-S-(2-aminoethyl)-L-cysteine

Supplier: Matrix Scientific

041785-1G 041785-5G
101853-178EA 547.69 USD
101853-178 101853-180
Di-fmoc-S-(2-aminoethyl)-L-cysteine
Di-fmoc-S-(2-aminoethyl)-L-cysteine

Hazard: Irritant

Formula: C₃₅H₃₂N₂O₆S
MW: 608.72 g/mol
MDL Number: MFCD12547717
CAS Number: 1786824-21-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR