Di-fmoc-L-homocystine

Supplier: Matrix Scientific

041784-1G 041784-5G
101853-174EA 661.54 USD
101853-174 101853-176
Di-fmoc-L-homocystine
Di-fmoc-L-homocystine

Hazard: Irritant

Formula: C₃₈H₃₆N₂O₈S₂
MDL Number: MFCD02682547
CAS Number: 181370-86-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR