Di-fmoc-L-cystathionine

Supplier: Matrix Scientific

041783-100MG 041783-250MG
101853-170EA 623.08 USD
101853-170 101853-172
Di-fmoc-L-cystathionine
Di-fmoc-L-cystathionine

Hazard: Irritant

Formula: C₃₇H₃₄N₂O₈S
MW: 666.76 g/mol
MDL Number: MFCD04974557
CAS Number: 622832-89-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR