Di-fmoc-DL-cystathionine

Supplier: Matrix Scientific

041782-100MG 041782-250MG
101853-166EA 623.08 USD
101853-166 101853-168
Di-fmoc-DL-cystathionine
Di-fmoc-DL-cystathionine

Hazard: Irritant

Formula: C₃₇H₃₄N₂O₈S
MDL Number: MFCD04974559
CAS Number: 1452573-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR