Di-boc-seleno-L-cystine

Supplier: Matrix Scientific

041781-250MG 041781-1G
101853-162EA 823.08 USD
101853-162 101853-164
Di-boc-seleno-L-cystine
Di-boc-seleno-L-cystine

Hazard: Irritant

Formula: C₁₆H₂₈N₂O₈Se₂
MW: 534.33 g/mol
MDL Number: MFCD04974560
CAS Number: 877754-71-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR