Di-boc-L-homocystine

Supplier: Matrix Scientific

041779-1G 041779-5G
101853-154EA 661.54 USD
101853-154 101853-156
Di-boc-L-homocystine
Di-boc-L-homocystine

Hazard: Irritant

Formula: C₁₈H₃₂N₂O₈S₂
Melting Pt: 152…154 °C
MDL Number: MFCD02682546
CAS Number: 130981-51-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR