Di-boc-DL-cystathionine

Supplier: Matrix Scientific

041777-100MG 041777-250MG
101853-146EA 623.08 USD
101853-146 101853-148
Di-boc-DL-cystathionine
Di-boc-DL-cystathionine

Hazard: Irritant

Formula: C₁₇H₃₀N₂O₈S
MW: 422.5 g/mol
MDL Number: MFCD04974558
CAS Number: 1452575-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR