Boc-S-trityl-L-penicillamine

Supplier: Matrix Scientific

Synonyms: Boc-Pen(Trt)-OH

041776-1G 041776-5G
101853-142EA 498.46 USD
101853-142 101853-144
Boc-S-trityl-L-penicillamine
Boc-S-trityl-L-penicillamine

Hazard: Irritant

Formula: C₂₉H₃₃NO₄S
MW: 491.646 g/mol
MDL Number: MFCD00237381
CAS Number: 135592-13-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR