Boc-S-trityl-D-penicillamine

Supplier: Matrix Scientific

Synonyms: Boc-D-Pen(Trt)-OH

041774-1G 041774-5G
101853-134EA 498.46 USD
101853-134 101853-136
Boc-S-trityl-D-penicillamine
Boc-S-trityl-D-penicillamine

Hazard: Irritant

Formula: C₂₉H₃₃NO₄S
MDL Number: MFCD00237382
CAS Number: 135592-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR