Boc-S-t-butyl-D-cysteine

Supplier: Matrix Scientific

041772-1G 041772-5G
101853-126EA 249.23 USD
101853-126 101853-128
Boc-S-t-butyl-D-cysteine
Boc-S-t-butyl-D-cysteine

Hazard: Irritant

Formula: C₁₂H₂₃NO₄S
MDL Number: MFCD02682528
CAS Number: 872211-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR