N-Boc-S-methyl-L-cysteine

Supplier: Matrix Scientific

Synonyms: Boc-Cys(Me)-OH

041771-1G 041771-5G
101853-122EA 124.62 USD
101853-122 101853-124
N-Boc-S-methyl-L-cysteine
N-Boc-S-methyl-L-cysteine

Hazard: Irritant

Formula: C₉H₁₇NO₄S
MW: 235.298 g/mol
MDL Number: MFCD00076923
CAS Number: 16947-80-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR