Boc-Cys(Et)-OH

Supplier: Matrix Scientific

Synonyms: (R)-2-((tert-Butoxycarbonyl)amino)-3-(ethylthio)propanoic acid

041770-1G 041770-5G
101853-118EA 138.46 USD
101853-118 101853-120
Boc-Cys(Et)-OH
Boc-Cys(Et)-OH

Hazard: Irritant

Formula: C₁₀H₁₉NO₄S
MW: 249.325 g/mol
MDL Number: MFCD00076919
CAS Number: 16947-82-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR