Boc-S-carboxymethyl-L-cysteine

Supplier: Matrix Scientific

041768-1G 041768-5G
101853-110EA 275.38 USD
101853-110 101853-112
Boc-S-carboxymethyl-L-cysteine
Boc-S-carboxymethyl-L-cysteine

Hazard: Irritant

Formula: C₁₀H₁₇NO₆S
MDL Number: MFCD02682533
CAS Number: 1354485-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR