Boc-S-carboxyethyl-L-cysteine

Supplier: Matrix Scientific

041767-1G 041767-5G
101853-106EA 326.15 USD
101853-106 101853-108
Boc-S-carboxyethyl-L-cysteine
Boc-S-carboxyethyl-L-cysteine

Hazard: Irritant

Formula: C₁₁H₁₉NO₆S
MW: 293.34 g/mol
MDL Number: MFCD02682531
CAS Number: 1354484-98-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR