Boc-S-carbamoyl-L-cysteine

Supplier: Matrix Scientific

041766-1G 041766-5G
101853-102EA 326.15 USD
101853-102 101853-104
Boc-S-carbamoyl-L-cysteine
Boc-S-carbamoyl-L-cysteine

Hazard: Irritant

Formula: C₉H₁₆N₂O₅S
MW: 264.3 g/mol
MDL Number: MFCD02682529
CAS Number: 1354487-65-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR