(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid

Supplier: Matrix Scientific

Synonyms: Boc-Cys(Bzl)-OH, N-Boc-S-benzyl-L-cysteine

041765-5G 041765-25G
101853-098EA 40 USD
101853-098 101853-100
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid
(R)-3-(Benzylthio)-2-((tert-butoxycarbonyl)amino)propanoic acid

Hazard: Irritant

Formula: C₁₅H₂₁NO₄S
MDL Number: MFCD00065567
CAS Number: 5068-28-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR