Boc-S-benzyl-DL-homocysteine

Supplier: Matrix Scientific

041764-250MG 041764-1G
101853-094EA 498.46 USD
101853-094 101853-096
Boc-S-benzyl-DL-homocysteine
Boc-S-benzyl-DL-homocysteine

Hazard: Irritant

Formula: C₁₆H₂₃NO₄S
MDL Number: MFCD02667646
CAS Number: 1290053-06-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR