Boc-Pen(ACM)-OH
Supplier: Matrix Scientific
Synonyms:
Boc-Pen(Acm)-OH
041762-1G
041762-5G
101853-086EA
300
USD
101853-086
101853-088
Boc-Pen(ACM)-OH
Boc-Pen(ACM)-OH
Hazard: Irritant
Formula:
C₁₃H₂₄N₂O₅S MW: 320.404 g/mol |
CAS Number:
129972-45-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...