Boc-S-acetamidomethyl-D-penicillamine

Supplier: Matrix Scientific

Synonyms: Boc-D-Pen(Acm)-OH

041761-1G 041761-5G
101853-082EA 326.15 USD
101853-082 101853-084
Boc-S-acetamidomethyl-D-penicillamine
Boc-S-acetamidomethyl-D-penicillamine

Hazard: Irritant

Formula: C₁₃H₂₄N₂O₅S
MW: 320.41 g/mol
MDL Number: MFCD00151884
CAS Number: 201421-14-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR