Boc-D-Pen(pMeBzl)-OH . DCHA

Supplier: Matrix Scientific

041759-1G 041759-5G
101853-074EA 363.08 USD
101853-074 101853-076
Boc-D-Pen(pMeBzl)-OH . DCHA
Boc-D-Pen(pMeBzl)-OH . DCHA

Hazard: Irritant

Formula: C₁₈H₂₇NO₄S·C₁₂H₂₃N
MDL Number: MFCD00235908
CAS Number: 198470-36-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR