Boc-S-[2-(4-pyridyl)ethyl]-L-cysteine

Supplier: Matrix Scientific

041756-1G 041756-5G
101853-062EA 735.38 USD
101853-062 101853-064
Boc-S-[2-(4-pyridyl)ethyl]-L-cysteine
Boc-S-[2-(4-pyridyl)ethyl]-L-cysteine

Hazard: Irritant

Formula: C₁₅H₂₂N₂O₄S
MDL Number: MFCD02682562
CAS Number: 151545-15-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR