Boc-S-[2-(4-pyridyl)ethyl]-DL-penicillamine

Supplier: Matrix Scientific

041755-100MG 041755-250MG
101853-058EA 412.31 USD
101853-058 101853-060
Boc-S-[2-(4-pyridyl)ethyl]-DL-penicillamine
Boc-S-[2-(4-pyridyl)ethyl]-DL-penicillamine
Formula: C₁₇H₂₆N₂O₄S
MDL Number: MFCD02682564
CAS Number: 1219318-32-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR