Boc-L-selenomethionine
Supplier: Matrix Scientific
041754-500MG
041754-1G
101853-054EA
686.15
USD
101853-054
101853-056
Boc-L-selenomethionine
Boc-L-selenomethionine
Hazard: Irritant
Formula:
C₁₀H₁₉NO₄Se |
MDL Number:
MFCD04974401 CAS Number: 45172-44-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...