Boc-DL-selenomethionine

Supplier: Matrix Scientific

041750-500MG 041750-1G
101853-034EA 823.08 USD
101853-034 101853-036
Boc-DL-selenomethionine
Boc-DL-selenomethionine

Hazard: Irritant

Formula: C₁₀H₁₉NO₄Se
MDL Number: MFCD02682568
CAS Number: 1219410-27-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR