Boc-DL-ethionine

Supplier: Matrix Scientific

041748-1G 041748-5G
101853-026EA 498.46 USD
101853-026 101853-028
Boc-DL-ethionine
Boc-DL-ethionine

Hazard: Irritant

Formula: C₁₁H₂₁NO₄S
MW: 263.36 g/mol
MDL Number: MFCD09750515
CAS Number: 16947-91-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR