Boc-DL-buthionine

Supplier: Matrix Scientific

041746-1G 041746-5G
101853-018EA 661.54 USD
101853-018 101853-020
Boc-DL-buthionine
Boc-DL-buthionine

Hazard: Irritant

Formula: C₁₃H₂₅NO₄S
MDL Number: MFCD12547714
CAS Number: 1396969-20-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR