Boc-Cys(3-(Boc-amino)propyl)-OH

Supplier: Matrix Scientific

041744-250MG 041744-1G 041744-5G
101853-006EA 300 USD
101853-006 101853-008 101853-010
Boc-Cys(3-(Boc-amino)propyl)-OH
Boc-Cys(3-(Boc-amino)propyl)-OH

Hazard: Irritant

Formula: C₁₆H₃₀N₂O₆S
MDL Number: MFCD09750514
CAS Number: 1446694-43-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR