Boc-α-methyl-DL-methionine

Supplier: Matrix Scientific

041742-1G 041742-5G
101852-996EA 958.46 USD
101852-996 101852-998
Boc-α-methyl-DL-methionine
Boc-alpha-methyl-DL-methionine

Hazard: Irritant

Formula: C₁₁H₂₁NO₄S
MW: 263.36 g/mol
MDL Number: MFCD02682556
CAS Number: 75691-21-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR