Ac-Cys(farnesyl)-OH

Supplier: Matrix Scientific

041740-100MG 041740-500MG
101852-988EA 420 USD
101852-988 101852-990
Ac-Cys(farnesyl)-OH
Ac-Cys(farnesyl)-OH

Hazard: Irritant

Formula: C₂₀H₃₃NO₃S
MDL Number: MFCD00871540
CAS Number: 135304-07-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR