Fmoc-5-amino-2-methoxybenzoic acid

Supplier: Matrix Scientific

041739-1G 041739-5G
101852-984EA 213.85 USD
101852-984 101852-986
Fmoc-5-amino-2-methoxybenzoic acid
Fmoc-5-amino-2-methoxybenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₅
MDL Number: MFCD04972603
CAS Number: 165073-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR