Fmoc-5-acetamido-2-aminobenzoic acid

Supplier: Matrix Scientific

041737-1G 041737-5G
101852-976EA 1730.77 USD
101852-976 101852-978
Fmoc-5-acetamido-2-aminobenzoic acid
Fmoc-5-acetamido-2-aminobenzoic acid

Hazard: Irritant

Formula: C₂₄H₂₀N₂O₅
MW: 416.44 g/mol
MDL Number: MFCD02682144
CAS Number: 1185296-79-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR