Fmoc-4-methylaminobenzoic acid

Supplier: Matrix Scientific

041736-1G 041736-5G
101852-972EA 224.62 USD
101852-972 101852-974
Fmoc-4-methylaminobenzoic acid
Fmoc-4-methylaminobenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MDL Number: MFCD04972272
CAS Number: 160977-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR