4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid
Supplier: Matrix Scientific
Synonyms:
Fmoc-4-Amb-OH, 4-(Fmoc-aminomethyl)benzoic Acid
041735-1G
041735-5G
101852-968EA
50.77
USD
101852-968
101852-970
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid
4-[[(9H-Fluoren-9-ylmethoxy)carbonyl]aminomethyl]benzoic acid
Hazard: Irritant
Formula:
C₂₃H₁₉NO₄ MW: 373.41 g/mol |
MDL Number:
MFCD01074693 CAS Number: 164470-64-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...