Fmoc-4-Abz-OH

Supplier: Matrix Scientific

041734-1G 041734-5G
101852-964EA 16.92 USD
101852-964 101852-966
Fmoc-4-Abz-OH
Fmoc-4-Abz-OH

Hazard: Irritant

Formula: C₂₂H₁₇NO₄
MDL Number: MFCD00144888
CAS Number: 185116-43-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR