Fmoc-4-amino-3-methylbenzoic acid

Supplier: Matrix Scientific

041733-1G 041733-5G
101852-960EA 498.46 USD
101852-960 101852-962
Fmoc-4-amino-3-methylbenzoic acid
Fmoc-4-amino-3-methylbenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MDL Number: MFCD02682226
CAS Number: 892878-63-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR