Fmoc-4-amino-3-methoxybenzoic acid

Supplier: Matrix Scientific

041732-1G 041732-5G
101852-956EA 547.69 USD
101852-956 101852-958
Fmoc-4-amino-3-methoxybenzoic acid
Fmoc-4-amino-3-methoxybenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₅
MDL Number: MFCD02682218
CAS Number: 254737-60-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR