Fmoc-Mamb-OH
Supplier: Matrix Scientific
Synonyms:
Fmoc-3-Amb-OH
041730-1G
041730-5G
101852-948EA
224.62
USD
101852-948
101852-950
Fmoc-Mamb-OH
Fmoc-Mamb-OH
Hazard: Irritant
Formula:
C₂₃H₁₉NO₄ |
MDL Number:
MFCD00273463 CAS Number: 155369-11-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...