Fmoc-3-aminobenzene-1,2-dicarboxylic acid
Supplier: Matrix Scientific
041728-1G
041728-5G
101852-940EA
326.15
USD
101852-940
101852-942
Fmoc-3-aminobenzene-1,2-dicarboxylic acid
Fmoc-3-aminobenzene-1,2-dicarboxylic acid
Hazard: Irritant
Formula:
C₂₃H₁₇NO₆ |
MDL Number:
MFCD02682148 CAS Number: 1185300-97-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...