Fmoc-3-amino-4-methylbenzoic acid

Supplier: Matrix Scientific

041727-1G 041727-5G
101852-936EA 224.62 USD
101852-936 101852-938
Fmoc-3-amino-4-methylbenzoic acid
Fmoc-3-amino-4-methylbenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MDL Number: MFCD01861379
CAS Number: 1072901-59-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR