Fmoc-3-amino-4-methoxybenzoic acid
Supplier: Matrix Scientific
Synonyms:
3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-methoxybenzoic acid
041726-1G
041726-5G
101852-932EA
55.38
USD
101852-932
101852-934
Fmoc-3-amino-4-methoxybenzoic acid
Fmoc-3-amino-4-methoxybenzoic acid
Hazard: Irritant
Formula:
C₂₃H₁₉NO₅ MW: 389.41 g/mol |
MDL Number:
MFCD02682216 CAS Number: 256935-69-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...