Fmoc-3-amino-4-chlorobenzoic acid

Supplier: Matrix Scientific

041725-1G 041725-5G
101852-928EA 158.46 USD
101852-928 101852-930
Fmoc-3-amino-4-chlorobenzoic acid
Fmoc-3-amino-4-chlorobenzoic acid

Hazard: Irritant

Formula: C₂₂H₁₆ClNO₄
MDL Number: MFCD02682163
CAS Number: 186320-11-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR