Fmoc-3-amino-2-methylbenzoic acid

Supplier: Matrix Scientific

041724-1G 041724-5G
101852-924EA 264.62 USD
101852-924 101852-926
Fmoc-3-amino-2-methylbenzoic acid
Fmoc-3-amino-2-methylbenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MDL Number: MFCD01861304
CAS Number: 1072901-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR