Fmoc-N-methylaminobenzoic acid

Supplier: Matrix Scientific

041723-1G 041723-5G
101852-920EA 187.69 USD
101852-920 101852-922
Fmoc-N-methylaminobenzoic acid
Fmoc-N-methylaminobenzoic acid

Hazard: Irritant

Formula: C₂₃H₁₉NO₄
MDL Number: MFCD04972602
CAS Number: 120467-46-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR