Fmoc-2-aminobenzene-1,4-dicarboxylic acid

Supplier: Matrix Scientific

041721-1G 041721-5G
101852-912EA 326.15 USD
101852-912 101852-914
Fmoc-2-aminobenzene-1,4-dicarboxylic acid
Fmoc-2-aminobenzene-1,4-dicarboxylic acid

Hazard: Irritant

Formula: C₂₃H₁₇NO₆
MDL Number: MFCD02682146
CAS Number: 1185298-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR