Fmoc-2-amino-5-methylbenzoic acid
Supplier: Matrix Scientific
041717-1G
041717-5G
101852-896EA
498.46
USD
101852-896
101852-898
Fmoc-2-amino-5-methylbenzoic acid
Fmoc-2-amino-5-methylbenzoic acid
Hazard: Irritant
Formula:
C₂₃H₁₉NO₄ |
MDL Number:
MFCD02682222 CAS Number: 332121-91-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...