Fmoc-2-amino-5-methoxybenzoic acid
Supplier: Matrix Scientific
041716-1G
041716-5G
101852-892EA
686.15
USD
101852-892
101852-894
Fmoc-2-amino-5-methoxybenzoic acid
Fmoc-2-amino-5-methoxybenzoic acid
Hazard: Irritant
Formula:
C₂₃H₁₉NO₅ MW: 389.41 g/mol |
MDL Number:
MFCD02682215 CAS Number: 332121-93-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...