Fmoc-2-amino-5-fluorobenzoic acid

Supplier: Matrix Scientific

041714-1G 041714-5G
101852-884EA 536.92 USD
101852-884 101852-886
Fmoc-2-amino-5-fluorobenzoic acid
Fmoc-2-amino-5-fluorobenzoic acid

Hazard: Irritant

Formula: C₂₂H₁₆FNO₄
MW: 377.38 g/mol
MDL Number: MFCD02682191
CAS Number: 1185296-60-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR