Fmoc-2-amino-5-bromobenzoic acid

Supplier: Matrix Scientific

041712-1G 041712-5G
101852-876EA 326.15 USD
101852-876 101852-878
Fmoc-2-amino-5-bromobenzoic acid
Fmoc-2-amino-5-bromobenzoic acid

Hazard: Irritant

Formula: C₂₂H₁₆BrNO₄
MW: 438.29 g/mol
MDL Number: MFCD02682151
CAS Number: 183871-04-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR