Fmoc-2-amino-4-fluorobenzoic acid

Supplier: Matrix Scientific

041711-1G 041711-5G
101852-872EA 536.92 USD
101852-872 101852-874
Fmoc-2-amino-4-fluorobenzoic acid
Fmoc-2-amino-4-fluorobenzoic acid

Hazard: Irritant

Formula: C₂₂H₁₆FNO₄
MDL Number: MFCD02682189
CAS Number: 1185298-00-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR