Fmoc-2-amino-4-chlorobenzoic acid

Supplier: Matrix Scientific

041710-1G 041710-5G
101852-868EA 326.15 USD
101852-868 101852-870
Fmoc-2-amino-4-chlorobenzoic acid
Fmoc-2-amino-4-chlorobenzoic acid

Hazard: Irritant

Formula: C₂₂H₁₆ClNO₄
MDL Number: MFCD02682157
CAS Number: 332121-92-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR